Learn More
Medchemexpress LLC 5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrinato iron(III) chloride | 90384-82-0 | 99.0% | 1024.27 g/mol | C44H28ClFeN4O12S4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1316975MG
Encompass_Preferred
FeTPPS (5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrinato iron(III) chloride) is a water-soluble ferric porphyrin complex used in biochemical research as a peroxynitrite decomposition catalyst and peroxynitrite scavenger. It is applied to study oxidative and nitrosative stress, modulate nitric oxide-related pathways, and evaluate cytoprotective effects against reactive oxygen species. Analytical-grade material with high purity suitable for in vitro and in vivo experiments.
  • Water-soluble ferric porphyrin complex.
  • Scavenges peroxynitrite and catalyzes its decomposition.
  • Reduces nitric oxide-mediated nitration in biological systems.
  • Used in studies of oxidative and nitrosative stress.
  • High purity for analytical and experimental applications.
  • Available in small-mass packages suitable for laboratory assays.
Catalog No. 50-002-62818