Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Aminoethylamino)-1-Naphthalene sulfonic acid, MP Biomedicals™

Catalog No. icn15474001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15474001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15474001 Supplier MP Biomedicals, Inc Supplier No. 0215474001
Add to Cart
Add to Cart

Synthetic. Soluble in DMSO and 1N NaOH (2.0mg/mL). Free Acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50402-56-7
Molecular Formula C12H14N2O3S
Molecular Weight (g/mol) 266.32
MDL Number MFCD00066932
InChI Key SJQRQOKXQKVJGJ-UHFFFAOYSA-N
Synonym 5-2-aminoethyl amino naphthalene-1-sulfonic acid, 1,5-edans, 5-2-aminoethylamino-1-naphthalenesulfonic acid, edans, 5-2-aminoethylamino-1-naphthalene sulfonic acid, 5-2-aminoethylamino naphthalene-1-sulfonic acid, n-aminoethyl-5-naphthylamine-1-sulfonic acid, 1-naphthalenesulfonicacid, 5-2-aminoethyl amino
PubChem CID 92329
IUPAC Name 5-[(2-aminoethyl)amino]naphthalene-1-sulfonic acid
SMILES NCCNC1=C2C=CC=C(C2=CC=C1)S(O)(=O)=O

Specifications

Color Gray-Yellow
Quantity 1 g
Formula Weight 266.32
Physical Form Crystalline Powder
Chemical Name or Material 5-(2-Aminoethylamino)-1-Naphthalene sulfonic acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.