Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt, 97%

Catalog No. AAL0655503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0655503 1 g
AAL06555MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0655503 Supplier Thermo Scientific Chemicals Supplier No. L0655503
Add to Cart

CAS: 100900-07-0 | C12H13N2NaO3S | 288.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100900-07-0
Molecular Formula C12H13N2NaO3S
Molecular Weight (g/mol) 288.297
MDL Number MFCD00051474
InChI Key HGWRACRQRUQQGH-UHFFFAOYSA-M
Synonym sodium 5-2-aminoethyl amino naphthalene-1-sulfonate, 5-2-aminoethylamino-1-naphthalenesulfonic acid sodium salt, sodium 5-2-aminoethylamino naphthalene-1-sulfonate, n-aminoethyl-5-naphthylamine-1-sulfonic acid sodium salt, 5-2-aminoethylamino-1-naphthalenesulphonic acid sodium salt, 1,5-edans, sodium salt, sodium 5-2-aminoethylamino-1-naphthalenesulfonate, sodium n-2-aminoethyl-5-naphthylamine-1-sulfonate, sodium5-2-aminoethylamino-1-naphthalenesulfonatehydrate
PubChem CID 4116252
IUPAC Name sodium;5-(2-aminoethylamino)naphthalene-1-sulfonate
SMILES C1=CC2=C(C=CC=C2S(=O)(=O)[O-])C(=C1)NCCN.[Na+]

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 4283850
Formula Weight 288.3
Percent Purity 97%
Chemical Name or Material 5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.