Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Bromophenyl)-3-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™

Catalog No. CC49610DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC49610DA 1 g
CC49610CB 250 mg
CC49610DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC49610DA Supplier Thermo Scientific Chemicals Supplier No. CC49610DA
Add to Cart

Chemical Identifiers

CAS 375857-64-0
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
InChI Key NYAFKUCVAVUCHW-UHFFFAOYSA-N
PubChem CID 18001310
IUPAC Name 5-(2-bromophenyl)-3-methyl-1,2,4-oxadiazole
SMILES CC1=NOC(=N1)C2=CC=CC=C2Br

Specifications

Assay Percent Range 97%
Quantity 1 g
Synonym 5-2-bromophenyl-3-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-2-bromophenyl-3-methyl, 5-2-bromophenyl-3-methyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,5-2-bromophenyl-3-methyl
Formula Weight 239.07
Percent Purity 97%
Chemical Name or Material 5-(2-Bromophenyl)-3-methyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.