Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC18203DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC18203DA 1 g
CC18203CB 250 mg
CC18203DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC18203DA Supplier Thermo Scientific Chemicals Supplier No. CC18203DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 215434-25-6
Molecular Formula C8H6ClNO2S3
Molecular Weight (g/mol) 279.771
InChI Key YIKNRCVJBOLZFL-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonyl chloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-sulphonyl chloride, 5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulphonylchloride, 5-2-methyl-thiazol-4-yl-thiophene-2-sulfonyl chloride, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenesulfonyl chloride, chloro 5-2-methyl 1,3-thiazol-4-yl 2-thienyl sulfone
PubChem CID 2736979
IUPAC Name 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride
SMILES CC1=NC(=CS1)C2=CC=C(S2)S(=O)(=O)Cl

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 279.79
Percent Purity 95%
Chemical Name or Material 5-(2-Methyl-1, 3-thiazol-4-yl)thiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.