Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(3-Bromophenyl)isoxazole, Thermo Scientific™

Catalog No. AAH5058003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5058003 1 g
AAH50580MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5058003 Supplier Thermo Scientific Chemicals Supplier No. H5058003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7064-33-7
Molecular Formula C31H38N4O4S2
Molecular Weight (g/mol) 594.79
MDL Number MFCD02183531
InChI Key ALUPLTWVMIXEQH-DICXZTSXNA-N
Synonym 5-3-bromophenyl isoxazole, 5-3-bromophenyl-1,2-oxazole, isoxazole,5-3-bromophenyl, zlchem 1341, acmc-209ofi
PubChem CID 2735607
IUPAC Name 5-(3-bromophenyl)-1,2-oxazole
SMILES CCCN1C(=O)C(C#N)=C(C)C(\C=C2/SC(=S)N(CCC3=CC=C(OC)C(OC)=C3)C2=O)=C1N1CC(C)CC(C)C1

Specifications

Quantity 1 g
Formula Weight 224.06
Chemical Name or Material 5-(3-Bromophenyl)isoxazole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.