Learn More
Accela Chembio Inc 5-(3-bromophenyl)oxazole | 5g | 243455-57-4 | MFCD05668970 | 97+% | Shelf Life: 2520 Days | Light Sensitive
Supplier:  Accela Chembio Inc SY0035065G
Encompass_Preferred
5-(3-bromophenyl)oxazole | 5g | 243455-57-4 | MFCD05668970 | 97+% | Shelf Life: 2520 Days | Light Sensitive
Catalog No. 50-250-165
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.