Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-(3-Pyridinyl)-1,3,4-thiadiazol-2-amine

Catalog No. CDS002566 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002566 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002566 Supplier Sigma Aldrich Supplier No. CDS00256650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-methoxytetrahydropyran is methoxy-substituted tetrahydropyran
  • α- and α-glycosidic c1-o1 bonds of the axial and equatorial forms of 2-methoxytetrahydropyran has been investigated by ab initio conformational study
  • Stereochemical properties of the glycosidic linkage of 2-methoxytetrahydropyran have been studied by the quantum-chemical pcilo method
  • It undergoes elimination reaction in the gas phase to yield 3, 4-dihydro-2h-pyran and methanol
  • Proton resonance spectra of 2-methoxytetrahydropyran has been analyzed

Specifications

CAS 68787-52-0
Molecular Formula C7H6N4S
Linear Formula C7H6N4S
Quantity 25 g
MDL Number MFCD00469439
Molecular Weight (g/mol) 178.21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.