Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole 96.0+%, TCI America™
SDP

Catalog No. T35121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35121G 1 g
T35125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35121G Supplier TCI America Supplier No. T35121G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 837392-64-0
Molecular Formula C14H18BNO3
Molecular Weight (g/mol) 259.112
MDL Number MFCD08669935
InChI Key BXFPTCYBFJOZHJ-UHFFFAOYSA-N
Synonym 5-Oxindoleboronic Acid Pinacol Ester, 2-(Oxindole-5-yl)-4,4,5,5-Tetramethyl-1,3,2-dioxaborolane, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-2-one
PubChem CID 21931486
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-dihydroindol-2-one
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)NC(=O)C3

Specifications

Melting Point 234°C
Quantity 1 g
Formula Weight 259.11
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)oxindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.