Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Dimethyl-2,4-oxazolidinedione 99.0+%, TCI America™
SDP

Catalog No. D000325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D000325G 25 g
D00035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D000325G Supplier TCI America Supplier No. D000325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 695-53-4
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00005379
InChI Key JYJFNDQBESEHJQ-UHFFFAOYSA-N
Synonym dimethadione, 5,5-dimethyloxazolidine-2,4-dione, 5,5-dimethyl-2,4-oxazolidinedione, propazone, dimethadion, eupractone, dimethyloxazolidinedione, 2,4-oxazolidinedione, 5,5-dimethyl, 5,5-dimethyloxazolidinedione, dimetadione dcit
PubChem CID 3081
IUPAC Name 5,5-dimethyl-1,3-oxazolidine-2,4-dione
SMILES CC1(C)OC(=O)NC1=O

Specifications

Melting Point 79°C
Color White-Yellow
Boiling Point 137°C
Quantity 25 g
Formula Weight 129.12
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,5-Dimethyl-2,4-oxazolidinedione
EINECSNumber (5)-0180
RTECSNumber RP9100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.