Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Dimethylhexane-2,4-dione, 99%

Catalog No. p-7046091
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1310403 1 g
AAL1310406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1310403 Supplier Thermo Scientific Chemicals Supplier No. L1310403
Only null left
Add to Cart

CAS: 7307-04-2 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7307-04-2
MDL Number MFCD00142767

Specifications

Boiling Point 60°C (10 mmHg)
Flash Point 57°C (134°F)
Molecular Formula C8H14O2
Refractive Index 1.46
Quantity 1 g
UN Number UN1224
Beilstein 969813
Synonym 2,2-dimethyl-3,5-hexanedione, 5,5-dimethyl-2,4-hexanedione, pivaloylacetone, keto form, 2,4-hexanedione, 5,5-dimethyl, 2,2-dimethylhexane-3,5-dione, pivaloylacetone, acmc-1bc1d, 2,2-dimethylhexan-3,5-dione, 5,5-dimethyl-hexan-2,4-dione, 5,5-dimethyl-2,4-hexanedione #
InChI Key LCLCVVVHIPPHCG-UHFFFAOYSA-N
SMILES CC(=O)CC(=O)C(C)(C)C
IUPAC Name 5,5-dimethylhexane-2,4-dione
Molecular Weight (g/mol) 142.20
PubChem CID 81741
Formula Weight 142.2
Percent Purity 99%
Chemical Name or Material 5,5-Dimethylhexane-2,4-dione
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 230-760-0
TSCA No

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.