Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Dimethylhydantoin, 97%

Catalog No. p-3757507
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116312500 250 g Plastic bottle
AC116310010 1 kg Plastic bottle
AC116310051 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC116312500 Supplier Thermo Scientific Chemicals Supplier No. 116312500
Only null left
Add to Cart

CAS: 77-71-4 | C5H8N2O2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-71-4
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00005266
InChI Key YIROYDNZEPTFOL-UHFFFAOYSA-N
Synonym 5,5-dimethylhydantoin, dimethylhydantoin, dm hydantoin, dantoin dmh, 2,4-imidazolidinedione, 5,5-dimethyl, 5,5-dimethyl-2,4-imidazolidinedione, dantoin 736, hydantoin, 5,5-dimethyl, 5,5-dimethylhydantoin dmh, 5,5-dimethyl hydantoin
PubChem CID 6491
IUPAC Name 5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C)NC(=O)NC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 175.0°C to 180.0°C
Color White
Loss on Drying 0.25% max. (60°C, 2 hrs, 1mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Quantity 250 g
Beilstein 24, 289
Fieser 07,126
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol and ether,soluble in ethanol,ethylether,aceton
Formula Weight 128.13
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 5, 5-Dimethylhydantoin
Show More Show Less
EINECSNumber 201-051-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.