Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Diphenylhydantoin 99.0+%, TCI America™
SDP

Catalog No. D0894500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0894500G 500 g
D089425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0894500G Supplier TCI America Supplier No. D0894500G

Chemical Identifiers

CAS 57-41-0
Molecular Formula C15H12N2O2
Molecular Weight (g/mol) 252.273
MDL Number MFCD00005264
InChI Key CXOFVDLJLONNDW-UHFFFAOYSA-N
Synonym phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan
PubChem CID 1775
ChEBI CHEBI:8107
IUPAC Name 5,5-diphenylimidazolidine-2,4-dione
SMILES C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3

Specifications

Melting Point 298°C
Color White
Quantity 500 g
Formula Weight 252.27
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5,5-Diphenylhydantoin
EINECSNumber (9)-0621
RTECSNumber MU1050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.