Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6,7,8-Tetrahydroquinoxaline 98.0+%, TCI America™
SDP

Catalog No. T140325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T140325G 25 g
T14035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T140325G Supplier TCI America Supplier No. T140325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34413-35-9
Molecular Formula C8H10N2
Molecular Weight (g/mol) 134.182
MDL Number MFCD00038078
InChI Key XCZPDOCRSYZOBI-UHFFFAOYSA-N
Synonym cyclohexapyrazine, quinoxaline, 5,6,7,8-tetrahydro, unii-m153cj9ra0, fema no. 3321, 5.6.7.8-tetrahydroquinoxaline, nutty quinoxaline, pubchem15475, acmc-209i7e, dsstox_cid_27678, dsstox_rid_82495
PubChem CID 36822
IUPAC Name 5,6,7,8-tetrahydroquinoxaline
SMILES C1CCC2=NC=CN=C2C1

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5,6,7,8-Tetrahydroquinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.