Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 5(6)-Aminofluorescein, Mixture Of Isomers, >95% Purity, 2 g

Catalog No. 502705278
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5278 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5278 Supplier Abcam Supplier No. AB1453432G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescein derivative with >95% purity for the preparation of isothiocyanates for fluorescent labeling.

  • Molecular weight: 347.32 g/mol
  • Purity: >95%
  • Composition: 100% 5(6)-Aminofluorescein (mixture of isomers)
  • Application: Fluorescein derivative typically used for the preparation of the isothiocyanate for fluorescent labeling

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Buffer 100% 5(6)-Aminofluorescein (mixture of isomers)
Detection Method Fluorescence
Formulation 100% 5(6)-Aminofluorescein (mixture of isomers)
Quantity 2 g
For Use With (Application) Fluorescent labeling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.