Learn More
Abcam 5(6)-Aminofluorescein, Mixture Of Isomers, Fluorescein Derivative, 10 g
Description
- Molecular Weight: 347.32 g/mol
- Purity: >95%
- Composition: 100% 5(6)-Aminofluorescein (mixture of isomers)
- CAS Number: 27599-63-9
- Storage Temperature: Store at Ambient
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Form | Powder |
| Shipping Condition | Ambient |
| Buffer | 100% 5(6)-Aminofluorescein (mixture of isomers) |
| Content And Storage | Store at ambient temperature. |
| Detection Method | Fluorescence |
| Formulation | 100% 5(6)-Aminofluorescein (mixture of isomers) |
| Quantity | 10 g |
| For Use With (Application) | Fluorescent labeling |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.