Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Diamino-1-methyluracil 98.0+%, TCI America™
SDP

Catalog No. D50101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50101G 1 g
D50105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50101G Supplier TCI America Supplier No. D50101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6972-82-3
Molecular Formula C5H8N4O2
Molecular Weight (g/mol) 156.145
MDL Number MFCD00091924
InChI Key PSIJQVXIJHUQPJ-UHFFFAOYSA-N
PubChem CID 157184
IUPAC Name 5,6-diamino-1-methylpyrimidine-2,4-dione
SMILES CN1C(=C(C(=O)NC1=O)N)N

Specifications

Melting Point 270°C
Color Yellow
Quantity 1 g
Formula Weight 156.15
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Diamino-1-methyluracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.