Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dichloroindole 98.0+%, TCI America™
SDP

Catalog No. D49241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49241G 1 g
D49245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49241G Supplier TCI America Supplier No. D49241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 121859-57-2
Molecular Formula C8H5Cl2N
Molecular Weight (g/mol) 186.04
MDL Number MFCD01075036
InChI Key ILINOHVVKWYAFM-UHFFFAOYSA-N
PubChem CID 10487776
IUPAC Name 5,6-dichloro-1H-indole
SMILES ClC1=C(Cl)C=C2C=CNC2=C1

Specifications

Melting Point 152°C
Quantity 1 g
Formula Weight 186.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichloroindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.