Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam 5(6)-FITC, Amine Reactive Label, >90% Purity, 100 mg
Description
- Molecular weight: 389.38 g/mol
- Purity: >90%
- Properties: Green fluorescent, amine-reactive label
- Applications: Fluorescence microscopy, flow cytometry, western blotting, and ELISA
- Function: Labels antibodies and proteins at primary amines
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Primary Amines |
| Label or Dye | FITC |
| Product Type | Protein Labeling Reagent |
| Buffer | 100% 3',6'-dihydroxy-6-isothiocyanato-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one |
| Content And Storage | Store at -20°C. |
| Form | Powder |
| Detection Method | Fluorescence |
| Quantity | 100 mg |
| For Use With (Application) | fluorescence microscopy, flow cytometry, western blotting, ELISA |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?