Learn More
Abcam 5(6)-FITC, Fluorescent Amine Reactive Label, Powder, 1 g
Description
- Molecular weight: 389.38 g/mol
- Purity: >90%
- Form: Powder
- Description: Green fluorescent, amine-reactive label for labeling antibodies and other purified proteins at primary amines (lysine side chains).
- Applications: Fluorescence microscopy, flow cytometry, western blotting and ELISA.
- Composition: 5-FITC (ab145325) and 6-FITC (ab146082) isomer mixture.
- Storage: Store at -20°C.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Proteins (Primary Amines) |
| Shipping Condition | Ambient |
| Label or Dye | FITC |
| Product Type | Protein Labeling Reagent |
| Buffer | 100% 3',6'-dihydroxy-6-isothiocyanato-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one |
| Content And Storage | Store at -20°C. |
| Form | Powder |
| Detection Method | Fluorescence |
| Formulation | 100% 3',6'-dihydroxy-6-isothiocyanato-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.