Learn More
Medchemexpress LLC L-ascorbic acid, 5,6-O-(1-methylethylidene) | 15042-01-0 | MFCD00010552 | 99.0% | 216.19 g/mol | C9H12O6 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01414110G
Encompass_Preferred
5,6-O-Isopropylidene-L-ascorbic acid is a protected derivative of L-ascorbic acid (ascorbic acid 5,6-acetonide) used as a research reagent in biochemical assays and synthetic chemistry. It is provided in solid and solution formats, characterized by high purity and defined storage recommendations to maintain stability.
  • High purity (99.0%) for reproducible experimental results.
  • Molecular weight 216.19 g/mol and formula C9H12O6 for accurate dosing and calculations.
  • Available as powder and 10 mM solution in DMSO for flexible workflows.
  • Stable under recommended storage conditions for extended shelf life.
  • Coa and sds documentation available to support quality and safety compliance.
Catalog No. 50-003-04185