Learn More
Medchemexpress LLC 5,6-O-isopropylidene-L-ascorbic acid | 15042-01-0 | MFCD00010552 | 99.0% | 216.19 g/mol | C9H12O6 | 5 G
Supplier: Medchemexpress LLC HYW0141415G
5,6-O-Isopropylidene-L-ascorbic acid is a protected derivative of L-ascorbic acid provided for research use. As an acetonide-protected form of vitamin C, it enables selective chemical transformations and serves as a reagent or intermediate in synthetic and biochemical applications.
- Protected derivative for selective synthetic transformations.
- Useful intermediate in vitamin C derivative synthesis.
- Retains antioxidant-related properties of ascorbic acid.
- High purity suitable for research applications.
- Available as solid and in ready-to-use solution formats.
- Stable as a powder under recommended low-temperature storage.