Learn More
AdipoGen 5 7-Dihydroxytryptamine HBr
SDP
Supplier:  AdipoGen CDXH0026M025
Encompass_Preferred
Chemical. CAS 31363-74-3 free base. Formula C10H12N2O2 . HBr. MW 192.2 . 80.9. Synthetic Autofluorescent serotonin derivative. Serotonergic neurotoxin, depleting serotonin and norepinephrine. Can be used for the identification of living serotonergic neurons even in the presence of dopaminergic neurons. Its selectively uptaken into 5-hydroxytryptamine neurons. It’s oxidation products are often more potent then the parent product.
Catalog No. 50-256-0795
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.