Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,7-Dimethylisatin, 96%

Catalog No. AAL1133206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1133206 5 g
AAL1133214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1133206 Supplier Thermo Scientific Chemicals Supplier No. L1133206
Add to Cart

CAS: 39603-24-2 | C10H9NO2 | 175.187 g/mol

5,7-Dimethylisatin is used to produce cyano-(5,7-dimethyl-2-oxo-1,2-dihydro-indol-3-ylidene)-acetic acid ethyl ester by knoevenagel condensation with cyanoacetic acid ethyl ester. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,7-Dimethylisatin is used to produce cyano-(5,7-dimethyl-2-oxo-1,2-dihydro-indol-3-ylidene)-acetic acid ethyl ester by knoevenagel condensation with cyanoacetic acid ethyl ester. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39603-24-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00047219
InChI Key HFZSCCJTJGWTDZ-UHFFFAOYSA-N
Synonym 5,7-dimethylisatin, 5,7-dimethylindoline-2,3-dione, 5,7-dimethyl-indole-2,3-dione, 5,7-dimethyl-2,3-dihydro-1h-indole-2,3-dione, 1h-indole-2,3-dione, 5,7-dimethyl, indole-2,3-dione, 5,7-dimethyl, 1h-indole-2,3-dione, 5,7-dimethyl-9ci, acmc-20a4zq, isatin-based compound, 48, 1h-indole-2,3-dione,5,7-dimethyl
PubChem CID 38296
IUPAC Name 5,7-dimethyl-1H-indole-2,3-dione
SMILES CC1=CC(=C2C(=C1)C(=O)C(=O)N2)C

Specifications

Melting Point 244°C to 248°C
Quantity 5 g
Beilstein 143679
Solubility Information Insoluble in water.
Formula Weight 175.19
Percent Purity 96%
Chemical Name or Material 5,7-Dimethylisatin
EINECSNumber 000-000-0
RTECSNumber NL7930000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.