Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,8-Dihydroxy-1,4-naphthoquinone 75.0+%, TCI America™
SDP

Catalog No. D20705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20705G 5 g
D20701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20705G Supplier TCI America Supplier No. D20705G

Chemical Identifiers

CAS 475-38-7
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.15
MDL Number MFCD00001685
InChI Key RQNVIKXOOKXAJQ-UHFFFAOYSA-N
Synonym naphthazarin, 5,8-dihydroxy-1,4-naphthoquinone, naphthazarine, naphthazarone, 5,8-dihydroxynaphthoquinone, 5,8-dihydroxy-1,4-naphthalenedione, naphthazalin, 5,8-dihydroxy-1,4-naphthosemiquinone, ccris 6670, dihydroxynaphthoquinone
PubChem CID 10141
ChEBI CHEBI:28849
IUPAC Name 5,8-dihydroxy-1,4-dihydronaphthalene-1,4-dione
SMILES OC1=CC=C(O)C2=C1C(=O)C=CC2=O

Specifications

Melting Point 230°C
Quantity 5 g
Formula Weight 190.15
Percent Purity ≥75.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,8-Dihydroxy-1,4-naphthoquinone
RTECSNumber QL7970000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.