Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Acetoacetamido)-2-benzimidazolinone 98.0+%, TCI America™
SDP

Catalog No. A1949250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1949250G 250 g
A194925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1949250G Supplier TCI America Supplier No. A1949250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26576-46-5
Molecular Formula C11H11N3O3
MDL Number MFCD00444508

Specifications

Color White-Yellow
Quantity 250 g
Formula Weight 233.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Acetoacetamido)-2-benzimidazolinone
EINECSNumber (5)-3506
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.