Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine 98.0+%, TCI America™
SDP

Catalog No. A209325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A209325G 25 g
A20935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A209325G Supplier TCI America Supplier No. A209325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25961-11-9
Molecular Formula C16H14ClNO
Molecular Weight (g/mol) 271.744
MDL Number MFCD01632170
InChI Key NMZOSOMVILZBJL-UHFFFAOYSA-N
PubChem CID 117681
IUPAC Name 1-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)ethanone
SMILES CC(=O)N1C2=CC=CC=C2CCC3=C1C=C(C=C3)Cl

Specifications

Melting Point 124°C
Color Yellow
Quantity 25 g
Synonym 5-Acetyl-3-chloro-2,2′-iminodibenzyl
Formula Weight 271.74
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.