Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetylindane, 97%

Catalog No. AAA1910209
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1910209 10 g
AAA1910288 2.5 g
AAA1910218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1910209 Supplier Thermo Scientific Chemicals Supplier No. A1910209
Add to Cart

CAS: 4228-10-8 | C11H12O | 160.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4228-10-8
MDL Number MFCD00021246

Specifications

Boiling Point 85°C to 87°C (0.05 mmHg)
Molecular Formula C11H12O
Quantity 10 g
Synonym 5-acetylindane, 5-acetylindan, 1-2,3-dihydro-1h-inden-5-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl ethan-1-one, ethanone,3-dihydro-1h-inden-5-yl, indan-5-ylethan-1-one, 1-indane-5-yl-ethanone, acmc-1arc8, 1-2,3-dihydro-1h-inden-6-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl-ethanone
InChI Key HCMWPTFPUCPKQM-UHFFFAOYSA-N
SMILES CC(=O)C1=CC2=C(CCC2)C=C1
IUPAC Name 1-(2,3-dihydro-1H-inden-5-yl)ethanone
Molecular Weight (g/mol) 160.22
PubChem CID 233976
Formula Weight 160.22
Percent Purity 97%
Chemical Name or Material 5-Acetylindane
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.