Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-methyl-3-phenyl-1H-pyrazole, 97%

Catalog No. AAA1575403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1575403 1 g
AAA1575406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1575403 Supplier Thermo Scientific Chemicals Supplier No. A1575403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 10199-50-5 | C10H11N3 | 173.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10199-50-5
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
MDL Number MFCD00067874
InChI Key KCYRMURRLLYLPU-UHFFFAOYSA-N
Synonym 1-methyl-3-phenyl-1h-pyrazol-5-amine, 5-amino-1-methyl-3-phenylpyrazole, 5-amino-1-methyl-3-phenyl-1h-pyrazole, 2-methyl-5-phenyl-2h-pyrazol-3-ylamine, 1-methyl-3-phenylpyrazole-5-ylamine, acmc-1bxci, maybridge1_004465, 1-methyl-3-phenyl-5-pyrazolamine, 1-methyl-3-phenyl-pyrazol-5-amine, 2-methyl-5-phenyl-pyrazol-3-amine
PubChem CID 517779
IUPAC Name 2-methyl-5-phenylpyrazol-3-amine
SMILES CN1C(=CC(=N1)C2=CC=CC=C2)N

Specifications

Melting Point 124°C to 130°C
Odor Odorless
Quantity 1 g
Beilstein 743807
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 5-Amino-1-methyl-3-phenyl-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.