Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-methyltetrazole 98.0+%, TCI America™
SDP

Catalog No. A242725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A242725G 25 g
A24275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A242725G Supplier TCI America Supplier No. A242725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5422-44-6
Molecular Formula C2H5N5
Molecular Weight (g/mol) 99.097
MDL Number MFCD00186454
InChI Key GTKOKCQMHAGFSM-UHFFFAOYSA-N
Synonym 1-Methyltetrazol-5-amine
PubChem CID 138492
IUPAC Name 1-methyltetrazol-5-amine
SMILES CN1C(=NN=N1)N

Specifications

Melting Point 228°C
Color White
Quantity 25 g
UN Number 1325
Formula Weight 99.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1-methyltetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.