Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-pentanol 97%, Thermo Scientific™

Catalog No. AC153830500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153830500 50 g
AC153832500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153830500 Supplier Thermo Scientific Chemicals Supplier No. 153830500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2508-29-4
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
InChI Key LQGKDMHENBFVRC-UHFFFAOYSA-N
Synonym 5-amino-1-pentanol, 5-aminopentanol, 1-pentanol, 5-amino, pentanol, 5-amino, 5-amino-1-pentanol in water, 5-amino pentanol, 5-amino-l-pentanol, 5-aminopentanol-1, 5-hydroxypentylamine, 1-amino-5-pentanol
PubChem CID 75634
IUPAC Name 5-aminopentan-1-ol
SMILES C(CCN)CCO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Brown
Infrared Spectrum Authentic
Refractive Index 1.4610 to 1.4630 (20°C, 589nm)
Linear Formula H2N(CH2)5OH
Quantity 50 g
Beilstein 04, I, 441
Formula Weight 103.16
Percent Purity ≥96.0%
Physical Form Liquid After Melting
Chemical Name or Material 5-Amino-1-pentanol, 97%
Show More Show Less
EINECSNumber 219-718-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.