Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1H-tetrazole 98.0+%, TCI America™
SDP

Catalog No. A0427100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0427100G 100 g
A042725G 25 g
A0427500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0427100G Supplier TCI America Supplier No. A0427100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4418-61-5
Molecular Formula CH3N5
Molecular Weight (g/mol) 85.07
MDL Number MFCD00005248
InChI Key ULRPISSMEBPJLN-UHFFFAOYSA-N
Synonym 5-aminotetrazole, 5-amino-1h-tetrazole, 1h-tetrazol-5-amine, aminotetrazole, tetrazol-5-ylamine, 1h-tetrazole, 5-amino, 5-amino-1,2,3,4-tetrazole, 5-amino tetrazole, 1h-tetrazol-5-ylamine, 5-amino-1h-tetrazol
PubChem CID 20467
IUPAC Name 2H-tetrazol-5-amine
SMILES C1(=NNN=N1)N

Specifications

Melting Point 201°C
Color White
Quantity 100 g
UN Number 1325
Formula Weight 85.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1H-tetrazole
EINECSNumber (5)-0627
RTECSNumber XF7465000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.