Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-chlorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A1913250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1913250G 250 g
A191325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1913250G Supplier TCI America Supplier No. A1913250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89-54-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007845
InChI Key GVCFFVPEOLCYNN-UHFFFAOYSA-N
Synonym 2-chloro-5-aminobenzoic acid, benzoic acid, 5-amino-2-chloro, 3-amino-6-chlorobenzoic acid, 5-amino-2-chloro-benzoic acid, 5-amino-2-chlorobenzoicacid, 3-carboxy-4-chloroaniline, 5-amino-2-chloro benzoic acid, zlchem 462, pubchem4654, acmc-209r1l
PubChem CID 37879
IUPAC Name 5-amino-2-chlorobenzoic acid
SMILES NC1=CC=C(Cl)C(=C1)C(O)=O

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 250 g
Formula Weight 171.58
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-chlorobenzoic Acid
EINECSNumber (3)-1480
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.