Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-chlorobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. C092225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C092225G 25 g
C09225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C092225G Supplier TCI America Supplier No. C092225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 320-51-4
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.569
MDL Number MFCD00007846
InChI Key ASPDJZINBYYZRU-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl aniline, 5-amino-2-chlorobenzotrifluoride, 3-trifluoromethyl-4-chloroaniline, 4-chloro-alpha,alpha,alpha-trifluoro-m-toluidine, 2-chloro-5-aminobenzotrifluoride, acbt, benzenamine, 4-chloro-3-trifluoromethyl, 2-chloro-5-aminebenzotrifluoride, 4-chloro-3-trifluoromethyl-phenylamine, 4-chloro-3-trifluoromethyl benzenamine
PubChem CID 67574
ChEBI CHEBI:59245
IUPAC Name 4-chloro-3-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1N)C(F)(F)F)Cl

Specifications

Quantity 25 g
UN Number 2811
Formula Weight 195.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 5-Amino-2-chlorobenzotrifluoride
EINECSNumber (3)-0266
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.