Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-mercaptobenzimidazole 97.0+%, TCI America™
SDP

Catalog No. A148225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A148225G 25 g
A14825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A148225G Supplier TCI America Supplier No. A148225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2818-66-8
Molecular Formula C7H7N3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00022671
InChI Key BXDMTLVCACMNJO-UHFFFAOYSA-N
Synonym 5-amino-2-mercaptobenzimidazole, 5-amino-2-benzimidazolethiol, 5-amino-1h-benzo d imidazole-2-thiol, 5-amino-1h-benzimidazole-2-thiol, 5-aminobenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-amino-1,3-dihydro, 2-benzimidazolethiol, 5-amino, 5-amino-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-amino benzimidazole, 5-amino-3h-1,3-benzodiazole-2-thiol
PubChem CID 2734002
IUPAC Name 5-amino-2,3-dihydro-1H-1,3-benzodiazole-2-thione
SMILES NC1=CC=C2NC(=S)NC2=C1

Specifications

Melting Point 237°C
Quantity 25 g
Formula Weight 165.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-mercaptobenzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.