Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methylbenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. A255225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A255225G 25 g
A25525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A255225G Supplier TCI America Supplier No. A255225G

Chemical Identifiers

CAS 6973-09-7
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD06681071
InChI Key KTPBKMYOIFHJMI-UHFFFAOYSA-N
Synonym 3-amino-6-methylbenzenesulfonamide, 2-methyl-5-aminobenzene sulfonamide, 5-amino-2-methylbenzene-1-sulfonamide, benzenesulfonamide, 5-amino-2-methyl, 5-amino-2-methyl-benzenesulfonamide, 3-aminosulfonyl-4-methylaniline, pubchem14390, p-aminotoluol-sulfonamid, acmc-1bcyh, ksc495k6b
PubChem CID 235511
IUPAC Name 5-amino-2-methylbenzenesulfonamide
SMILES CC1=C(C=C(C=C1)N)S(=O)(=O)N

Specifications

Melting Point 169°C
Quantity 25 g
Formula Weight 186.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-methylbenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.