Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methylbenzothiazole, 99%

Catalog No. p-7027121
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2363104 2 g
AAB2363109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2363104 Supplier Thermo Scientific Chemicals Supplier No. B2363104
Add to Cart

CAS: 13382-43-9 | C8H8N2S | 164.23 g/mol

5-Amino-2-methylbenzothiazole is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2-methylbenzothiazole is used as pharmaceutical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13382-43-9
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00226291
InChI Key GPWQHYMVUZYWIK-UHFFFAOYSA-N
Synonym 2-methylbenzo d thiazol-5-amine, 5-amino-2-methylbenzothiazole, 5-benzothiazolamine, 2-methyl, 2-methyl-5-benzothiazolamine, 2-methyl-5-aminobenzothiazole, 2-methylbenzothiazole-5-ylamine, 2-methyl-1,3-benzothiazol-5-ylamine, acmc-209v5w, cambridge id 5107842, 2-methylbenzothiazol-5-amine
PubChem CID 36229
IUPAC Name 2-methyl-1,3-benzothiazol-5-amine
SMILES CC1=NC2=CC(N)=CC=C2S1

Specifications

Quantity 2 g
Formula Weight 164.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Amino-2-methylbenzothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement

H301-H315-H319-H335

Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.