Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-methylindole 98.0+%, TCI America™
SDP

Catalog No. A15391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15391G 1 g
A15395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15391G Supplier TCI America Supplier No. A15391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7570-49-2
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD02093426
InChI Key JQULCCZIXYRBSE-UHFFFAOYSA-N
Synonym 5-amino-2-methylindole, 2-methyl-5-aminoindole, 2-methylindole-5-ylamine, pubchem7273, acmc-209ozq, 2-methylindole-5-amine, 2-methyl-5-amino-1h-indole, bidd:gt0193, 5-amino-2-methyl-1h-indole, 2-methyl-1h-indol-5-ylamine
PubChem CID 2733992
IUPAC Name 2-methyl-1H-indol-5-amine
SMILES CC1=CC2=CC(N)=CC=C2N1

Specifications

Quantity 1 g
Formula Weight 146.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-methylindole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.