Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-(methylthio)pyrimidine, 96%, Thermo Scientific™

Catalog No. AC449820010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449820010 1 g
AC449822500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449820010 Supplier Thermo Scientific Chemicals Supplier No. 449820010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 42382-46-7
Molecular Formula C5H7N3S
Molecular Weight (g/mol) 141.19
MDL Number MFCD10697095
InChI Key NRJNAXLXHFQYEX-UHFFFAOYSA-N
Synonym 5-amino-2-methylthio pyrimidine, 2-methylthio pyrimidin-5-amine, 2-methylsulfanyl pyrimidin-5-amine, 2-methylmercapto-5-aminopyrimidine, 5-pyrimidinamine,2-methylthio, 2-methylsulfanylpyrimidin-5-ylamine, 2-methylsulfanyl-5-pyrimidinamine, 2-methylthio-5-pyrimidinamine, 5-pyrimidinamine, 2-methylthio, 5-pyrimidinamine, 2-methylthio-9ci
PubChem CID 12215991
IUPAC Name 2-methylsulfanylpyrimidin-5-amine
SMILES CSC1=NC=C(N)C=N1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 141.19
Percent Purity 96%
Chemical Name or Material 5-Amino-2-(methylthio)pyrimidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.