Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-3-bromo-2-chloropyridine 95.0+%, TCI America™
SDP

Catalog No. A25391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25391G 1 g
A25395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25391G Supplier TCI America Supplier No. A25391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 130284-53-6
Molecular Formula C5H4BrClN2
Molecular Weight (g/mol) 207.46
MDL Number MFCD03840433
InChI Key ISKBXMMELYRESC-UHFFFAOYSA-N
PubChem CID 14595221
IUPAC Name 5-bromo-6-chloropyridin-3-amine
SMILES NC1=CC(Br)=C(Cl)N=C1

Specifications

Melting Point 113°C
Quantity 1 g
Formula Weight 207.46
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-3-bromo-2-chloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.