Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-6-chloro-2-picoline, 98+%

Catalog No. p-3777247
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC360740010 1 g Glass bottle
AC360740050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC360740010 Supplier Thermo Scientific Chemicals Supplier No. 360740010
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 39745-40-9 | C6H7ClN2 | 142.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39745-40-9
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD03095213
InChI Key SUMSBUDMYMYTLL-UHFFFAOYSA-N
Synonym 3-amino-2-chloro-6-picoline, 3-amino-2-chloro-6-methylpyridine, 5-amino-6-chloro-2-picoline, 2-chloro-6-methyl-pyridin-3-ylamine, 2-chloro-3-amino-6-methylpyridine, 3-pyridinamine, 2-chloro-6-methyl, 2-chloro-6-methyl-3-pyridylamine, pubchem1245, acmc-20dld6, 2-chloro-3-amino-6-picoline
PubChem CID 2734415
IUPAC Name 2-chloro-6-methylpyridin-3-amine
SMILES CC1=CC=C(N)C(Cl)=N1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 74°C to 80°C
Color Cream to Tan
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 142.59
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-6-chloro-2-picoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.