Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Azaindole, 98%

Catalog No. AAH6427214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6427214 25 g
AAH6427203 1 g
AAH6427206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6427214 Supplier Thermo Scientific Chemicals Supplier No. H6427214

CAS: 271-34-1 | C7H6N2 | 118.139 g/mol

5-Azaindole is a factor VIIa inhibitor. Also useful intermediate for pharmaceutical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Azaindole is a factor VIIa inhibitor. Also useful intermediate for pharmaceutical application.

Solubility
Hardly soluble in water. Soluble in methanol and chloroform.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 271-34-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00955936
InChI Key SRSKXJVMVSSSHB-UHFFFAOYSA-N
Synonym 5-azaindole, 1h-pyrrolo 3,2-c pyridine, 1,5-diazaindene, 3,7-dideazapurine, 5-azindole, 5h-pyrrolo 3,2-c pyridine, pyrrolo 3,2-c pyridine, 5-aza indole, 5-aza-indole
PubChem CID 9220
IUPAC Name 1H-pyrrolo[3,2-c]pyridine
SMILES C1=CNC2=C1C=NC=C2

Specifications

Melting Point 104°C to 108°C
Quantity 25 g
Beilstein 109684
Solubility Information Hardly soluble in water. Soluble in methanol and chloroform.
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 5-Azaindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UY8725000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.