Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Benzoylpentanoic Acid 98.0+%, TCI America™
SDP

Catalog No. B218225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B218225G 25 g
B21825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B218225G Supplier TCI America Supplier No. B218225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4144-62-1
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00014380
InChI Key AIEMSTCGCMIJTI-UHFFFAOYSA-N
Synonym 5-benzoylpentanoic acid, 5-benzoylvaleric acid, delta-benzoylvaleric acid, 5-benzoylpentanoicacid, 6-benzoylpentanoic acid, acmc-1akf8, 6-oxo-6-phenyl-hexanoic acid, 6-oxo-6-phenyl hexanoic acid, 6-keto-6-phenyl-hexanoic acid
PubChem CID 223595
IUPAC Name 6-oxo-6-phenylhexanoic acid
SMILES OC(=O)CCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 77°C
Color White
Quantity 25 g
Formula Weight 206.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzoylpentanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.