Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1-pentanol 90.0+%, TCI America™
SDP

Catalog No. B184825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B184825G 25 g
B18485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B184825G Supplier TCI America Supplier No. B184825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34626-51-2
Molecular Formula C5H11BrO
Molecular Weight (g/mol) 167.046
MDL Number MFCD00061101
InChI Key WJVQJXVMLRGNGA-UHFFFAOYSA-N
PubChem CID 118709
IUPAC Name 5-bromopentan-1-ol
SMILES C(CCO)CCBr

Specifications

Boiling Point 117°C
Quantity 25 g
Formula Weight 167.05
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-1-pentanol
EINECSNumber (2)-2002
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.