Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1-pentene 95.0+%, TCI America™
SDP

Catalog No. B147425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B147425G 25 g
B14745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B147425G Supplier TCI America Supplier No. B147425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1119-51-3
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.031
MDL Number MFCD00000264
InChI Key LPNANKDXVBMDKE-UHFFFAOYSA-N
Synonym 5-bromo-1-pentene, 1-pentene, 5-bromo, 1-bromo-4-pentene, 4-pentenyl bromide, 5-bromopentene, 5-bromo-pent-1-ene, 5-bromo-pentene, 5-bromopentene-1, 5-bromo-1-amylene, pent-4-enyl bromide
PubChem CID 70704
IUPAC Name 5-bromopent-1-ene
SMILES C=CCCCBr

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 g
UN Number 1993
Formula Weight 149.03
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-1-pentene
EINECSNumber (2)-0067
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.