Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2,3-pyridinediol 98.0+%, TCI America™
SDP

Catalog No. B278025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B278025G 25 g
B27805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B278025G Supplier TCI America Supplier No. B278025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34206-49-0
Molecular Formula C5H4BrNO2
Molecular Weight (g/mol) 190.00
MDL Number MFCD08276314
InChI Key KXBXUHWCODHRHK-UHFFFAOYSA-N
Synonym 5-Bromo-2,3-dihydroxypyridine
PubChem CID 12563083
IUPAC Name 5-bromo-3-hydroxy-1,2-dihydropyridin-2-one
SMILES OC1=CC(Br)=CNC1=O

Specifications

Melting Point 249°C
Color White-Yellow
Quantity 25 g
Formula Weight 190.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2,3-pyridinediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.