Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chloropyridine 98.0+%, TCI America™
SDP

Catalog No. B232925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B232925G 25 g
B23295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B232925G Supplier TCI America Supplier No. B232925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53939-30-3
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD01318951
InChI Key PEAOEIWYQVXZMB-UHFFFAOYSA-N
Synonym 2-chloro-5-bromopyridine, 5-bromo-2-chloro-pyridine, pyridine, 5-bromo-2-chloro, 2-chloro-5-bromo-pyridine, 2-chlorine-5-bromine pyridine, pubchem1168, acmc-1afhf, 3-bromo-6-chloropyridine, 2-chloro-5-bromo pyridine, 3-bromo-6-chloro-pyridine
PubChem CID 2734414
IUPAC Name 5-bromo-2-chloropyridine
SMILES C1=CC(=NC=C1Br)Cl

Specifications

Melting Point 70°C
Quantity 25 g
Formula Weight 192.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.