Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2'-Deoxyuridine, MP Biomedicals™

Catalog No. MP21001712 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21001712 250 mg
MP21001715 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP21001712 Supplier MP Biomedicals, Inc Supplier No. 02100171.2
Add to Cart
Add to Cart

Useful for the study of DNA synthesis. 5-Bromo-2'-Deoxyuridine, MP Biomedicals (5-BrdU) is a thymidine analogue that is incorporated into DNA. It is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently it is us

5-BrdU can be used to label DNA, which can be used in conjunction with BrdU Monoclonal Antibody for the immunohistochemical analysis of cell proliferation. Thymidine analog is used as a mutagen in genetic research. It is selectively incorporated into cellular DNA in place of thymidine during S-phase. It is also used to study cell signaling and other processes that induce cell proliferation.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-14-3
Molecular Formula C9H11BrN2O5
Molecular Weight (g/mol) 307.10
MDL Number MFCD00006529
InChI Key WOVKYSAHUYNSMH-RRKCRQDMSA-N
Synonym 5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine
PubChem CID 6035
ChEBI CHEBI:472552
IUPAC Name 5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O

Specifications

Melting Point 183°C to 193°C
Quantity 250 mg
Solubility Information Soluble in water (10mg/mL or 50–100mg/mL with heat) or sodium hydroxide (50mg/mL).
Formula Weight 307.1
Percent Purity ≥99%
Physical Form Powder
Recommended Storage −20°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.