Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-fluorophenol, 98%

Catalog No. AAH6151814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6151814 25 g
AAH6151806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6151814 Supplier Thermo Scientific Chemicals Supplier No. H6151814

CAS: 112204-58-7 | C6H4BrFO | 190.999 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112204-58-7
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD07782066
InChI Key YPTHSYKJDRMAJY-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromophenol, phenol, 5-bromo-2-fluoro, 2-fluoro-5-bromo phenol, 5-bromo-2-fluoro-phenol, 3-bromo-6-fluorophenol, 3-bromo-6-fluoro-phenol, acmc-2099dd, ksc493i2d
PubChem CID 183421
IUPAC Name 5-bromo-2-fluorophenol
SMILES C1=CC(=C(C=C1Br)O)F

Specifications

Color White
Boiling Point 53°C (0.2 mmHg)
Quantity 25 g
Formula Weight 191
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 5-Bromo-2-fluorophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.