Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-fluorophenol 98.0+%, TCI America™
SDP

Catalog No. B306425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B306425G 25 g
B30645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B306425G Supplier TCI America Supplier No. B306425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112204-58-7
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD07782066
InChI Key YPTHSYKJDRMAJY-UHFFFAOYSA-N
Synonym 2-fluoro-5-bromophenol, phenol, 5-bromo-2-fluoro, 2-fluoro-5-bromo phenol, 5-bromo-2-fluoro-phenol, 3-bromo-6-fluorophenol, 3-bromo-6-fluoro-phenol, acmc-2099dd, ksc493i2d
PubChem CID 183421
IUPAC Name 5-bromo-2-fluorophenol
SMILES C1=CC(=C(C=C1Br)O)F

Specifications

Color Yellow
Boiling Point 54°C
Quantity 25 g
Formula Weight 191.00
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-fluorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.